Product Details

SNP ID
rs142759394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101818248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTCATGGGACTCACCAACTCAC[A/G]GGAGATTCAGCTTCTACTTTTTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4F15 PubMed Links

Gene Details

Gene
OR4F15
Gene Name
olfactory receptor family 4 subfamily F member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001674.1 62 Missense Mutation CAG,CGG Q21R NP_001001674.1

View Full Product Details