Product Details

SNP ID
rs143038267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74902784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCCGGCTCACATCCTGGTTCAC[C/G]TGCTAAGAGAAGGAGAGGAGGGAAC
Phenotype
MIM: 154550
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM219B PubMed Links

Gene Details

Gene
FAM219B
Gene Name
family with sequence similarity 219 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321920.1 526 Missense Mutation CAC,CAG H144Q NP_001308849.1
NM_001321921.1 526 Missense Mutation CAC,CAG H144Q NP_001308850.1
NM_001321922.1 526 Intron NP_001308851.1
NM_001321923.1 526 Intron NP_001308852.1
NM_020447.4 526 Missense Mutation CAC,CAG H144Q NP_065180.1
XM_017022433.1 526 Intron XP_016877922.1
Gene
MPI
Gene Name
mannose phosphate isomerase
There are no transcripts associated with this gene.

View Full Product Details