Product Details

SNP ID
rs143616433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73321876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCACATGTGAGTGGCTATCTACA[C/T]GGCTCTACAGAGCGCTACATAGCTA
Phenotype
MIM: 605206
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCN4 PubMed Links

Gene Details

Gene
HCN4
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005477.2 5211 UTR 3 NP_005468.1
XM_011521148.2 5211 UTR 3 XP_011519450.1

View Full Product Details