Product Details
- SNP ID
-
rs144043304
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:74180114 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCACCATTGGCACCCAACAGGGCC[A/G]TCTTGCGGAAGACCTGCAGGGTTGG
- Phenotype
-
MIM: 602059
MIM: 610745
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ISLR
PubMed Links
Gene Details
- Gene
- ISLR
- Gene Name
- immunoglobulin superfamily containing leucine rich repeat
There are no transcripts associated with this gene.
- Gene
- STRA6
- Gene Name
- stimulated by retinoic acid 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001142617.1 |
2182 |
Missense Mutation |
ACG,ATG |
T657M |
NP_001136089.1 |
NM_001142618.1 |
2182 |
Missense Mutation |
ACG,ATG |
T657M |
NP_001136090.1 |
NM_001142619.1 |
2182 |
Missense Mutation |
ACG,ATG |
T648M |
NP_001136091.1 |
NM_001142620.1 |
2182 |
Intron |
|
|
NP_001136092.1 |
NM_001199040.1 |
2182 |
Missense Mutation |
ACG,ATG |
T694M |
NP_001185969.1 |
NM_001199041.1 |
2182 |
Missense Mutation |
ACG,ATG |
T672M |
NP_001185970.1 |
NM_001199042.1 |
2182 |
Missense Mutation |
ACG,ATG |
T696M |
NP_001185971.1 |
NM_022369.3 |
2182 |
Missense Mutation |
ACG,ATG |
T657M |
NP_071764.3 |
XM_011521883.1 |
2182 |
Missense Mutation |
ACG,ATG |
T657M |
XP_011520185.1 |
XM_011521884.1 |
2182 |
Missense Mutation |
ACG,ATG |
T594M |
XP_011520186.1 |
XM_011521885.2 |
2182 |
Intron |
|
|
XP_011520187.1 |
XM_017022478.1 |
2182 |
Missense Mutation |
ACG,ATG |
T673M |
XP_016877967.1 |
XM_017022479.1 |
2182 |
Missense Mutation |
ACG,ATG |
T657M |
XP_016877968.1 |
XM_017022480.1 |
2182 |
Missense Mutation |
ACG,ATG |
T594M |
XP_016877969.1 |
View Full Product Details