Product Details

SNP ID
rs144259269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50242423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAAAAGATTCTGACCCTGGAG[A/G]AGCCCCCATTCTTCACAGAGGCCTC
Phenotype
MIM: 142704 MIM: 603247
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDC PubMed Links

Gene Details

Gene
HDC
Gene Name
histidine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306146.1 2037 Missense Mutation TCC,TTC S576F NP_001293075.1
NM_002112.3 2037 Missense Mutation TCC,TTC S609F NP_002103.2
XM_017022094.1 2037 Missense Mutation TCC,TTC S644F XP_016877583.1
XM_017022095.1 2037 Missense Mutation TCC,TTC S611F XP_016877584.1
XM_017022096.1 2037 Missense Mutation TCC,TTC S568F XP_016877585.1
XM_017022097.1 2037 Missense Mutation TCC,TTC S565F XP_016877586.1
XM_017022098.1 2037 Missense Mutation TCC,TTC S500F XP_016877587.1
XM_017022099.1 2037 Intron XP_016877588.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
There are no transcripts associated with this gene.

View Full Product Details