Product Details

SNP ID
rs144669082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48826058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCGCTTCCAGAGGAGATGATTGG[C/T]AAACTGTTATCCATATGGTATCTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHC4 PubMed Links

Gene Details

Gene
SHC4
Gene Name
SHC adaptor protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203349.3 2235 Silent Mutation TTA,TTG L602L NP_976224.3
XM_005254375.3 2235 Silent Mutation TTA,TTG L419L XP_005254432.1

View Full Product Details