Product Details

SNP ID
rs144868755
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101818427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTGCTCAATTACAGCCCCTAAG[A/G]TGATTTGTGATATTTTCAAGAAGCA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR4F15 PubMed Links

Gene Details

Gene
OR4F15
Gene Name
olfactory receptor family 4 subfamily F member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001674.1 241 Missense Mutation ATG,GTG M81V NP_001001674.1

View Full Product Details