Product Details

SNP ID
rs144886450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45487808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCACTCTATCCTAGACAGTTTAC[A/G]TTACTTGCCTCATGGGACCTATTTC
Phenotype
MIM: 602095
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105376714 PubMed Links

Gene Details

Gene
LOC105376714
Gene Name
uncharacterized LOC105376714
There are no transcripts associated with this gene.

Gene
SLC30A4
Gene Name
solute carrier family 30 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321036.1 Intron NP_001307965.1
NM_013309.5 Intron NP_037441.2
XM_011521997.2 Intron XP_011520299.1
XM_017022560.1 Intron XP_016878049.1
XM_017022561.1 Intron XP_016878050.1
XM_017022562.1 Intron XP_016878051.1

View Full Product Details