Product Details

SNP ID
rs145281560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48825984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCAATACTGTCATTTGTTGGAA[G/T]GCAAAAGTGCTGGATTATTATCTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SHC4 PubMed Links

Gene Details

Gene
SHC4
Gene Name
SHC adaptor protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203349.3 2309 Missense Mutation CAT,CCT H627P NP_976224.3
XM_005254375.3 2309 Missense Mutation CAT,CCT H444P XP_005254432.1

View Full Product Details