Product Details

SNP ID
rs145762743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45587477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGGCCGTCGTCTCCGGACGGG[A/G]CCCTGACACGGCCACCCTACTGCCT
Phenotype
MIM: 604310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BLOC1S6 PubMed Links

Gene Details

Gene
BLOC1S6
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311255.1 355 Intron NP_001298184.1
NM_001311256.1 355 Intron NP_001298185.1
NM_012388.3 355 Missense Mutation ACC,GCC T12A NP_036520.1

View Full Product Details