Product Details

SNP ID
rs146901531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43234894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCTGTTGACAGGTACTGGCTGT[A/G]CTGGGAATAGGAGATTTTGCAGGGG
Phenotype
MIM: 177070 MIM: 603805
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPB42 PubMed Links

Gene Details

Gene
EPB42
Gene Name
erythrocyte membrane protein band 4.2
There are no transcripts associated with this gene.

Gene
TGM5
Gene Name
transglutaminase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004245.3 964 Missense Mutation CAC,TAC H502Y NP_004236.1
NM_201631.3 964 Missense Mutation CAC,TAC H584Y NP_963925.2
XM_011522230.2 964 Missense Mutation CAC,TAC H241Y XP_011520532.1
XM_017022729.1 964 Missense Mutation CAC,TAC H241Y XP_016878218.1

View Full Product Details