Product Details

SNP ID
rs147849550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50182625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCCTGGAGAAAGCGCGCCAGAC[C/G]CCACACAAGCCTTTTCTGCTCTTCC
Phenotype
MIM: 609123 MIM: 603247
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 430 Intron NP_079113.2
XM_011522046.2 430 Intron XP_011520348.1
XM_011522047.2 430 Intron XP_011520349.1
XM_011522048.1 430 Intron XP_011520350.1
XM_011522049.2 430 Intron XP_011520351.1
XM_011522051.2 430 Intron XP_011520353.1
XM_011522052.2 430 Intron XP_011520354.1
XM_011522053.1 430 Intron XP_011520355.1
XM_011522056.2 430 Intron XP_011520358.2
XM_011522058.2 430 Intron XP_011520360.1
XM_011522059.1 430 Intron XP_011520361.1
XM_011522060.1 430 Intron XP_011520362.1
XM_011522061.1 430 Intron XP_011520363.1
XM_011522062.1 430 Intron XP_011520364.1
XM_011522063.1 430 Intron XP_011520365.1
XM_011522064.1 430 Intron XP_011520366.1
XM_011522069.2 430 Intron XP_011520371.1
XM_011522070.1 430 Intron XP_011520372.1
XM_017022587.1 430 Intron XP_016878076.1
XM_017022588.1 430 Intron XP_016878077.1
XM_017022589.1 430 Intron XP_016878078.1
XM_017022590.1 430 Intron XP_016878079.1
XM_017022591.1 430 Intron XP_016878080.1
XM_017022592.1 430 Intron XP_016878081.1
XM_017022593.1 430 Intron XP_016878082.1
XM_017022594.1 430 Intron XP_016878083.1
XM_017022595.1 430 Intron XP_016878084.1
XM_017022596.1 430 Intron XP_016878085.1
XM_017022597.1 430 Intron XP_016878086.1
XM_017022598.1 430 Intron XP_016878087.1
XM_017022599.1 430 Intron XP_016878088.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159629.1 430 Silent Mutation ACC,ACG T66T NP_001153101.1
NM_003645.3 430 Silent Mutation ACC,ACG T66T NP_003636.2

View Full Product Details