Product Details

SNP ID
rs148981648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40701808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTTTTTTTTTTTTGAGAC[A/G]GAGTCTCGCTCTGTTGCCAGGCTGG
Phenotype
MIM: 179617
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD51 PubMed Links

Gene Details

Gene
RAD51
Gene Name
RAD51 recombinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164269.1 372 Silent Mutation ACA,ACG T76T NP_001157741.1
NM_001164270.1 372 Intron NP_001157742.1
NM_002875.4 372 Intron NP_002866.2
NM_133487.3 372 Silent Mutation ACA,ACG T76T NP_597994.3
XM_006720626.3 372 Intron XP_006720689.1
XM_011521857.2 372 Intron XP_011520159.1
XM_011521858.2 372 Intron XP_011520160.1
XM_011521859.2 372 Intron XP_011520161.1
XM_011521860.2 372 Intron XP_011520162.1
XM_011521861.2 372 Intron XP_011520163.1
XM_011521862.2 372 Intron XP_011520164.1
Gene
RAD51-AS1
Gene Name
RAD51 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details