Product Details

SNP ID
rs150067777
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79845222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTAATCCAGATTTAAGCTGTTGA[C/T]GAGTCTTCGTAAAGGACTTCATCTA
Phenotype
MIM: 604197
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MTHFS PubMed Links
Additional Information
For this assay, SNP(s) [rs8923] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MTHFS
Gene Name
5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199758.1 607 Silent Mutation TCA,TCG S143S NP_001186687.1
NM_006441.3 607 Silent Mutation TCA,TCG S200S NP_006432.1
Gene
ST20-MTHFS
Gene Name
ST20-MTHFS readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199760.1 607 Silent Mutation TCA,TCG S176S NP_001186689.1

View Full Product Details