Product Details

SNP ID
rs138160245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:90005161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGAGCTCAGGGCATGCCTGGG[G/T]CATCTCCAAAGCTCTGCTGAGAGTG
Phenotype
MIM: 603020
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AFG3L1P PubMed Links

Gene Details

Gene
AFG3L1P
Gene Name
AFG3 like matrix AAA peptidase subunit 1, pseudogene
There are no transcripts associated with this gene.

Gene
DBNDD1
Gene Name
dysbindin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042610.2 1785 UTR 3 NP_001036075.1
NM_001288708.1 1785 UTR 3 NP_001275637.1
NM_001288709.1 1785 UTR 3 NP_001275638.1
NM_024043.3 1785 UTR 3 NP_076948.2

View Full Product Details