Product Details

SNP ID
rs138895105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57473994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACGCGCTGGTAAATCTGCTCCCC[C/G]TCTTGTGTCTGGAAGGTATAGAGTC
Phenotype
MIM: 608333 MIM: 180663
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DOK4 PubMed Links

Gene Details

Gene
DOK4
Gene Name
docking protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018110.3 937 Silent Mutation GAC,GAG D215E NP_060580.2
XM_005256042.3 937 Missense Mutation GAC,GAG D215E XP_005256099.1
XM_005256043.2 937 Missense Mutation GAC,GAG D215E XP_005256100.1
Gene
POLR2C
Gene Name
RNA polymerase II subunit C
There are no transcripts associated with this gene.

View Full Product Details