Product Details
- SNP ID
-
rs138912985
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:67886145 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTCGGGAGCGCGGCACAGGCTC[C/T]GAAACCAACCAGGAGACGCTGCGGG
- Phenotype
-
MIM: 606030
MIM: 177015
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EDC4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73594554] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EDC4
- Gene Name
- enhancer of mRNA decapping 4
There are no transcripts associated with this gene.
- Gene
- NRN1L
- Gene Name
- neuritin 1 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198443.1 |
410 |
Silent Mutation |
TCC,TCT |
S128S |
NP_940845.1 |
- Gene
- PSKH1
- Gene Name
- protein serine kinase H1
There are no transcripts associated with this gene.
View Full Product Details