Product Details

SNP ID
rs139306010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66884415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTAAAATGGAGGCTCTTTTCCC[A/G]GGTGCCACCCACCCTAAACAGTTCC
Phenotype
MIM: 615499
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDP2 PubMed Links

Gene Details

Gene
PDP2
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005256064.4 460 Missense Mutation CAG,CGG Q44R XP_005256121.1
XM_005256066.4 460 Missense Mutation CAG,CGG Q44R XP_005256123.1
XM_005256067.4 460 Missense Mutation CAG,CGG Q44R XP_005256124.1
XM_005256068.4 460 Missense Mutation CAG,CGG Q44R XP_005256125.1
XM_017023506.1 460 Missense Mutation CAG,CGG Q44R XP_016878995.1

View Full Product Details