Product Details

SNP ID
rs140187592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:14672622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGGTAGTCAGTCACACTTGGG[C/T]GAGTCCGGCTCACATAGGAACTGGG
Phenotype
MIM: 603603
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BFAR PubMed Links

Gene Details

Gene
BFAR
Gene Name
bifunctional apoptosis regulator
There are no transcripts associated with this gene.

Gene
PLA2G10
Gene Name
phospholipase A2 group X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003561.2 827 Silent Mutation TCA,TCG S161S NP_003552.1
XM_011522695.2 827 Intron XP_011520997.1
XM_011522696.2 827 Intron XP_011520998.1
XM_011522697.1 827 Intron XP_011520999.1
XM_011522698.1 827 Intron XP_011521000.1

View Full Product Details