Product Details
- SNP ID
-
rs140464487
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:69715026 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAGCAAACTCTCCTATGAACACTC[A/G]CTCAAACCAGCCTTTCAGAATGGCA
- Phenotype
-
MIM: 125860
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NQO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11555215,rs200863930] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NQO1
- Gene Name
- NAD(P)H quinone dehydrogenase 1
View Full Product Details