Product Details
- SNP ID
-
rs140573852
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:30553830 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCAACTCCAGAAAACCTGAGTG[C/G]TACAGACCACTGTTTGCAAACTCAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF764
PubMed Links
Gene Details
- Gene
- ZNF764
- Gene Name
- zinc finger protein 764
View Full Product Details