Product Details

SNP ID
rs140988362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27067031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTCAGCTGCCCCCCATTTTTCT[C/T]GAGGGAGAAAAAGGGGAATCCTCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf82 PubMed Links

Gene Details

Gene
C16orf82
Gene Name
chromosome 16 open reading frame 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145545.1 134 Silent Mutation CTC,CTT L12L NP_001139017.1

View Full Product Details