Product Details

SNP ID
rs141195654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66884792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTCCCTGATGTCCCACCAGACC[C/T]TGGAGCACATGGAGGGAGCTATGGA
Phenotype
MIM: 615499
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDP2 PubMed Links

Gene Details

Gene
PDP2
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005256064.4 837 Silent Mutation CTG,TTG L170L XP_005256121.1
XM_005256066.4 837 Silent Mutation CTG,TTG L170L XP_005256123.1
XM_005256067.4 837 Silent Mutation CTG,TTG L170L XP_005256124.1
XM_005256068.4 837 Silent Mutation CTG,TTG L170L XP_005256125.1
XM_017023506.1 837 Silent Mutation CTG,TTG L170L XP_016878995.1

View Full Product Details