Product Details

SNP ID
rs141912338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1334710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCGGGCCCGCCAGGTGACCTG[C/T]ACCTGGGCACCGCCATCGGCTTCGC
Phenotype
MIM: 604009 MIM: 601661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAIAP3 PubMed Links

Gene Details

Gene
BAIAP3
Gene Name
BAI1 associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199096.1 702 Intron NP_001186025.1
NM_001199097.1 702 Intron NP_001186026.1
NM_001199098.1 702 Intron NP_001186027.1
NM_001199099.1 702 Intron NP_001186028.1
NM_001286464.1 702 Intron NP_001273393.1
NM_003933.4 702 Missense Mutation CAC,TAC H17Y NP_003924.2
XM_011522728.1 702 Missense Mutation CAC,TAC H17Y XP_011521030.1
XM_011522729.1 702 Missense Mutation CAC,TAC H17Y XP_011521031.1
XM_011522730.2 702 Missense Mutation CAC,TAC H17Y XP_011521032.1
Gene
UBE2I
Gene Name
ubiquitin conjugating enzyme E2 I
There are no transcripts associated with this gene.

View Full Product Details