Product Details
- SNP ID
-
rs142345157
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:4883402 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTGCCTACTTCTGCCCAGATAC[C/G]AAGACCGCCAGCTCCTGGATGGACA
- Phenotype
-
MIM: 602871
MIM: 609771
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PPL
PubMed Links
Gene Details
- Gene
- PPL
- Gene Name
- periplakin
- Gene
- UBN1
- Gene Name
- ubinuclein 1
There are no transcripts associated with this gene.
View Full Product Details