Product Details

SNP ID
rs142821011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71634359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTGATGGTGCTGACGGGGAGAA[A/G]GCCCAGCAACTGGATGTCCAGTTCT
Phenotype
MIM: 614094
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARVELD3 PubMed Links

Gene Details

Gene
MARVELD3
Gene Name
MARVEL domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017967.3 839 Intron NP_001017967.2
NM_001271329.1 839 Intron NP_001258258.1
NM_052858.5 839 Silent Mutation AAA,AAG K254K NP_443090.4
XM_011523449.2 839 Intron XP_011521751.1

View Full Product Details