Product Details

SNP ID
rs144020066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19502109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGGAAAGAGCAGTTTGGGGTCA[A/T]GGGCTCAACAATATAAAGGCAGATT
Phenotype
MIM: 605943
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GDE1 PubMed Links

Gene Details

Gene
GDE1
Gene Name
glycerophosphodiester phosphodiesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324066.1 2521 UTR 3 NP_001310995.1
NM_001324067.1 2521 UTR 3 NP_001310996.1
NM_016641.3 2521 UTR 3 NP_057725.1
Gene
TMC5
Gene Name
transmembrane channel like 5
There are no transcripts associated with this gene.

View Full Product Details