Product Details

SNP ID
rs144773756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:14685532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGTAAGTTTACCAGGTGTTGGT[G/T]GTAGGGAGGAATGGGAACGGATTGC
Phenotype
MIM: 603603
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G10 PubMed Links

Gene Details

Gene
PLA2G10
Gene Name
phospholipase A2 group X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003561.2 Intron NP_003552.1
XM_011522695.2 Intron XP_011520997.1
XM_011522696.2 Intron XP_011520998.1
XM_011522697.1 Intron XP_011520999.1
XM_011522698.1 Intron XP_011521000.1

View Full Product Details