Product Details

SNP ID
rs145224500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30555859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAGACGTGCTCCACCAGTGTGG[A/G]GCGCCAGGCAAAGCTCTTCCCGCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF764 PubMed Links

Gene Details

Gene
ZNF764
Gene Name
zinc finger protein 764
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172679.1 701 Missense Mutation CCC,TCC P187S NP_001166150.1
NM_033410.3 701 Missense Mutation CCC,TCC P188S NP_219363.2

View Full Product Details