Product Details

SNP ID
rs145423593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:51137316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGGAATTGGAGGGATGCCACC[A/G]TTCTGAATGACGGAGATCTCGTTGG
Phenotype
MIM: 602218
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105371253 PubMed Links

Gene Details

Gene
LOC105371253
Gene Name
uncharacterized LOC105371253
There are no transcripts associated with this gene.

Gene
SALL1
Gene Name
spalt like transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127892.1 3802 Silent Mutation AAC,AAT N1160N NP_001121364.1
NM_002968.2 3802 Silent Mutation AAC,AAT N1257N NP_002959.2
XM_006721241.3 3802 Silent Mutation AAC,AAT N1257N XP_006721304.1
XM_011523254.2 3802 Silent Mutation AAC,AAT N1257N XP_011521556.1

View Full Product Details