Product Details

SNP ID
rs146071407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67661095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCGATAGCATCGTCGAGGTGAA[A/G]AGCAAAGTAAGGGCTCCTCCGGCCT
Phenotype
MIM: 609377 MIM: 610859 MIM: 607484
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACD PubMed Links

Gene Details

Gene
ACD
Gene Name
adrenocortical dysplasia homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082486.1 140 Intron NP_001075955.1
NM_001082487.1 140 Intron NP_001075956.1
NM_022914.2 140 Intron NP_075065.2
XM_005256115.3 140 Intron XP_005256172.1
Gene
C16orf86
Gene Name
chromosome 16 open reading frame 86
There are no transcripts associated with this gene.

Gene
CARMIL2
Gene Name
capping protein regulator and myosin 1 linker 2
There are no transcripts associated with this gene.

Gene
ENKD1
Gene Name
enkurin domain containing 1
There are no transcripts associated with this gene.

Gene
PARD6A
Gene Name
par-6 family cell polarity regulator alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037281.1 140 Silent Mutation AAA,AAG K19K NP_001032358.1
NM_016948.2 140 Silent Mutation AAA,AAG K19K NP_058644.1
XM_005255977.3 140 Silent Mutation AAA,AAG K19K XP_005256034.1
XM_011523095.2 140 Silent Mutation AAA,AAG K19K XP_011521397.1
XM_011523096.2 140 Intron XP_011521398.1
XM_017023261.1 140 Intron XP_016878750.1

View Full Product Details