Product Details

SNP ID
rs146094039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30479455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGAAGAAACTCAGCAACACTTC[A/G]TACCAGGTAAAAAAGTTAGTTAGGA
Phenotype
MIM: 153370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGAL PubMed Links

Gene Details

Gene
ITGAL
Gene Name
integrin subunit alpha L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114380.1 746 Intron NP_001107852.1
NM_002209.2 746 Silent Mutation TCA,TCG S190S NP_002200.2
XM_005255313.1 746 Silent Mutation TCA,TCG S190S XP_005255370.1
XM_006721044.1 746 Intron XP_006721107.1
XM_011545849.1 746 Silent Mutation TCA,TCG S94S XP_011544151.1

View Full Product Details