Product Details

SNP ID
rs147086833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:685171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGTCGATGTAGGAAGTGTACCA[C/G]TGCTCCTGGGGGAGGGAGCGCCCGG
Phenotype
MIM: 609082 MIM: 603264 MIM: 607207
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXL16 PubMed Links

Gene Details

Gene
FBXL16
Gene Name
F-box and leucine rich repeat protein 16
There are no transcripts associated with this gene.

Gene
JMJD8
Gene Name
jumonji domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005920.2 3313 Intron NP_001005920.2
NM_001323918.1 3313 Intron NP_001310847.1
NM_001323919.1 3313 Intron NP_001310848.1
NM_001323920.1 3313 Intron NP_001310849.1
NM_001323922.1 3313 Intron NP_001310851.1
Gene
LOC105371184
Gene Name
uncharacterized LOC105371184
There are no transcripts associated with this gene.

Gene
RHBDL1
Gene Name
rhomboid like 1
There are no transcripts associated with this gene.

Gene
STUB1
Gene Name
STIP1 homology and U-box containing protein 1
There are no transcripts associated with this gene.

Gene
WDR24
Gene Name
WD repeat domain 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032259.3 3313 Missense Mutation CAC,CAG H675Q NP_115635.1
XM_011522699.2 3313 Missense Mutation CAC,CAG H764Q XP_011521001.1

View Full Product Details