Product Details

SNP ID
rs147817697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74873851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTCTGCACTGGCATCAAGAGAA[C/T]GCTGCTCGGTGGTTTAAGGCTAACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR59 PubMed Links

Gene Details

Gene
WDR59
Gene Name
WD repeat domain 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324171.1 3127 Intron NP_001311100.1
NM_001324172.1 3127 Intron NP_001311101.1
NM_030581.3 3127 UTR 3 NP_085058.3
XM_005256146.3 3127 UTR 3 XP_005256203.1
XM_011523331.2 3127 UTR 3 XP_011521633.1
XM_011523332.2 3127 UTR 3 XP_011521634.1
XM_017023667.1 3127 UTR 3 XP_016879156.1
XM_017023668.1 3127 UTR 3 XP_016879157.1
XM_017023669.1 3127 UTR 3 XP_016879158.1
XM_017023670.1 3127 UTR 3 XP_016879159.1

View Full Product Details