Product Details

SNP ID
rs147961036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4260516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTGGTGCAGCAGCCTCACCTGT[G/T]CCTGCTGCTGCTGCAGCTGCACCTG
Phenotype
MIM: 600743
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC01569 PubMed Links

Gene Details

Gene
LINC01569
Gene Name
long intergenic non-protein coding RNA 1569
There are no transcripts associated with this gene.

Gene
LOC105371063
Gene Name
putative uncharacterized protein encoded by LINC00596
There are no transcripts associated with this gene.

Gene
TFAP4
Gene Name
transcription factor AP-4 (activating enhancer binding protein 4)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003223.2 995 Missense Mutation GAA,GCA E202A NP_003214.1
XM_011522633.2 995 Missense Mutation GAA,GCA E189A XP_011520935.1
XM_011522635.2 995 Missense Mutation GAA,GCA E142A XP_011520937.1

View Full Product Details