Product Details

SNP ID
rs148033494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10531973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAGTTTTGGGGCCCTAGATGTA[G/T]ACAGCTGTGGCGATGGAGGCCAAGG
Phenotype
MIM: 602334
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EMP2 PubMed Links

Gene Details

Gene
EMP2
Gene Name
epithelial membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001424.5 1658 UTR 3 NP_001415.1
XM_006720864.3 1658 UTR 3 XP_006720927.1

View Full Product Details