Product Details

SNP ID
rs148244001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1827436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCGCGTACACTCGCAACCTGCA[C/T]CACGAGCTGGAGCTGGGCGTGGTGA
Phenotype
MIM: 616320 MIM: 138760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAHD1 PubMed Links

Gene Details

Gene
FAHD1
Gene Name
fumarylacetoacetate hydrolase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018104.2 213 Silent Mutation CAC,CAT H69H NP_001018114.1
NM_001142398.1 213 Silent Mutation CAC,CAT H69H NP_001135870.1
NM_031208.3 213 Intron NP_112485.1
Gene
HAGH
Gene Name
hydroxyacylglutathione hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040427.1 213 Intron NP_001035517.1
NM_001286249.1 213 Intron NP_001273178.1
NM_005326.4 213 Intron NP_005317.2
XM_011522469.2 213 Intron XP_011520771.1
XM_011522470.2 213 Intron XP_011520772.1
XM_017023195.1 213 Intron XP_016878684.1
Gene
MEIOB
Gene Name
meiosis specific with OB domains
There are no transcripts associated with this gene.

View Full Product Details