Product Details

SNP ID
rs149042359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68022809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGCAGCACACGGGGCTCCAGGC[A/C]CAGGTCAGCAAAGCTGCCCTTAGAC
Phenotype
MIM: 607618 MIM: 609707
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DDX28 PubMed Links

Gene Details

Gene
DDX28
Gene Name
DEAD-box helicase 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018380.3 1059 Missense Mutation GGC,TGC G132C NP_060850.2
Gene
DUS2
Gene Name
dihydrouridine synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271762.1 1059 Intron NP_001258691.1
NM_001271763.1 1059 Intron NP_001258692.1
NM_017803.4 1059 Intron NP_060273.1
Gene
LOC100131303
Gene Name
uncharacterized LOC100131303
There are no transcripts associated with this gene.

View Full Product Details