Product Details

SNP ID
rs149386661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71933972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCAGCAGAAAGCCCACCACCTC[A/G]TCCCAGGCTCGGCTCAGTGTCCTGC
Phenotype
MIM: 616434 MIM: 607895
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IST1 PubMed Links

Gene Details

Gene
IST1
Gene Name
IST1, ESCRT-III associated factor
There are no transcripts associated with this gene.

Gene
PKD1L3
Gene Name
polycystin 1 like 3, transient receptor potential channel interacting
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181536.1 4767 Silent Mutation GAC,GAT D1589D NP_853514.1
XM_017023201.1 4767 Intron XP_016878690.1
XM_017023202.1 4767 Intron XP_016878691.1
XM_017023203.1 4767 Intron XP_016878692.1
XM_017023204.1 4767 Intron XP_016878693.1

View Full Product Details