Product Details
- SNP ID
-
rs149593870
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3137625 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAGCACCCGGGAAGCGGTGAGGA[C/G]GCTGTCGCCTTGGTGGAGGACCTAC
- Phenotype
-
MIM: 608387
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CASP16P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7198126] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CASP16P
- Gene Name
- caspase 16, pseudogene
There are no transcripts associated with this gene.
- Gene
- ZNF213
- Gene Name
- zinc finger protein 213
- Gene
- ZNF213-AS1
- Gene Name
- ZNF213 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details