Product Details

SNP ID
rs149692669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69329134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGTGCTGGATGATGCGGGCTGCC[C/T]ACCCGCTCGCCTGCCACACCACCTG
Phenotype
MIM: 606979 MIM: 609982
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COG8 PubMed Links

Gene Details

Gene
COG8
Gene Name
component of oligomeric golgi complex 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032382.4 645 UTR 3 NP_115758.3
Gene
PDF
Gene Name
peptide deformylase (mitochondrial)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022341.1 645 Nonsense Mutation TAG,TGG *207W NP_071736.1
Gene
VPS4A
Gene Name
vacuolar protein sorting 4 homolog A
There are no transcripts associated with this gene.

View Full Product Details