Product Details

SNP ID
rs149781615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27268932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACGCCCGCTCTCGATCCTTAAT[A/G]ATGGGAAGGGGCTTCCACTTGTCGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLJ21408 PubMed Links

Gene Details

Gene
FLJ21408
Gene Name
uncharacterized LOC400512
There are no transcripts associated with this gene.

Gene
NSMCE1
Gene Name
NSE1 homolog, SMC5-SMC6 complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145080.3 Intron NP_659547.2
XM_006721023.3 Intron XP_006721086.1
XM_011545761.2 Intron XP_011544063.1
XM_011545762.1 Intron XP_011544064.1
XM_017023040.1 Intron XP_016878529.1
XM_017023041.1 Intron XP_016878530.1
XM_017023042.1 Intron XP_016878531.1

View Full Product Details