Product Details

SNP ID
rs149826573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85780438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTCGTACACGTGGATCGTAGG[C/T]GCTACGCAGTCCATCGTAAACTTGG
Phenotype
MIM: 604886
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMC8 PubMed Links

Gene Details

Gene
EMC8
Gene Name
ER membrane protein complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142288.1 352 Intron NP_001135760.1
NM_006067.4 352 Silent Mutation GCA,GCG A138A NP_006058.1
XM_011522813.2 352 Intron XP_011521115.1
XM_017022867.1 352 Silent Mutation GCA,GCG A86A XP_016878356.1

View Full Product Details