Product Details

SNP ID
rs150345189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30555880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAGCGCCAGGCAAAGCTCTTCC[C/T]GCACACGTAGCAGCCATGACGCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF764 PubMed Links

Gene Details

Gene
ZNF764
Gene Name
zinc finger protein 764
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172679.1 680 Missense Mutation AGG,GGG R180G NP_001166150.1
NM_033410.3 680 Missense Mutation AGG,GGG R181G NP_219363.2

View Full Product Details