Product Details

SNP ID
rs151037166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:54932605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGGGACCCAGCGTACCGGAAGAA[C/T]GCCACAAGGGACGCCACGGCTACCC
Phenotype
MIM: 615624 MIM: 606195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRNDE PubMed Links

Gene Details

Gene
CRNDE
Gene Name
colorectal neoplasia differentially expressed (non-protein coding)
There are no transcripts associated with this gene.

Gene
IRX5
Gene Name
iroquois homeobox 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252197.1 800 Silent Mutation AAC,AAT N119N NP_001239126.1
NM_005853.5 800 Silent Mutation AAC,AAT N119N NP_005844.4
XM_011522809.1 800 Silent Mutation AAC,AAT N49N XP_011521111.1

View Full Product Details