Product Details

SNP ID
rs151087701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84976480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATCCCTTCCCATCGCAGCCTCC[G/T]CTCCCATGTGGGCTTCTCACTTTTC
Phenotype
MIM: 614604
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZDHHC7 PubMed Links

Gene Details

Gene
ZDHHC7
Gene Name
zinc finger DHHC-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145548.1 654 Silent Mutation NP_001139020.1
NM_017740.2 654 Silent Mutation NP_060210.2
XM_011523222.1 654 Intron XP_011521524.1
XM_017023434.1 654 Missense Mutation XP_016878923.1
XM_017023435.1 654 Missense Mutation XP_016878924.1
XM_017023436.1 654 Missense Mutation XP_016878925.1

View Full Product Details