Product Details

SNP ID
rs151105071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75529504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTCCCGCGCCAGGTCCTCGAAG[C/G]GCACCAGGCGGTAGCGGCCGCGCAG
Phenotype
MIM: 604817 MIM: 614949
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHST5 PubMed Links

Gene Details

Gene
CHST5
Gene Name
carbohydrate sulfotransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024533.4 2276 Missense Mutation CAC,CGC H294R NP_078809.2
Gene
TMEM231
Gene Name
transmembrane protein 231
There are no transcripts associated with this gene.

View Full Product Details