Product Details

SNP ID
rs138026530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81822540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCAGCACACCCCAGAGCCAAGA[C/G]AGACCCAGCAGCAGCACCTCAGGGT
Phenotype
MIM: 138033 MIM: 616514
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GCGR PubMed Links

Gene Details

Gene
GCGR
Gene Name
glucagon receptor
There are no transcripts associated with this gene.

Gene
MCRIP1
Gene Name
MAPK regulated corepressor interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001093767.2 1004 UTR 3 NP_001087236.1
NM_001288798.1 1004 UTR 3 NP_001275727.1
NM_001288799.1 1004 UTR 3 NP_001275728.1
NM_207368.4 1004 UTR 3 NP_997251.2
XM_011523571.2 1004 UTR 3 XP_011521873.1
XM_011523572.2 1004 UTR 3 XP_011521874.1

View Full Product Details