Product Details

SNP ID
rs138537888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3724447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATCAGTGCCTCCCTGTTCAGCT[C/G]TCTGGCCTCGCCCTGCCCCGGGTCC
Phenotype
MIM: 609240 MIM: 604682
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GSG2 PubMed Links

Gene Details

Gene
GSG2
Gene Name
germ cell associated 2, haspin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031965.2 545 Missense Mutation TCT,TGT S171C NP_114171.2
Gene
ITGAE
Gene Name
integrin subunit alpha E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002208.4 545 Intron NP_002199.3
XM_011523823.2 545 Intron XP_011522125.1
XM_011523825.2 545 Intron XP_011522127.1
XM_011523827.2 545 Intron XP_011522129.1
XM_011523828.2 545 Intron XP_011522130.1
XM_017024586.1 545 Intron XP_016880075.1
XM_017024587.1 545 Intron XP_016880076.1

View Full Product Details