Product Details

SNP ID
rs138557340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44074760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTGTCTTCGCAGGTGGCCACT[C/T]GGGCCCGCAGGTATACCCTTGGCAT
Phenotype
MIM: 611045 MIM: 605315 MIM: 611793
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
G6PC3 PubMed Links

Gene Details

Gene
G6PC3
Gene Name
glucose-6-phosphatase catalytic subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319945.1 865 Missense Mutation CGG,TGG R136W NP_001306874.1
NM_138387.3 865 Missense Mutation CGG,TGG R136W NP_612396.1
XM_011525473.2 865 Missense Mutation CGG,TGG R21W XP_011523775.1
XM_011525474.2 865 Missense Mutation CGG,TGG R21W XP_011523776.1
XM_017025335.1 865 Missense Mutation CGG,TGG R21W XP_016880824.1
Gene
HDAC5
Gene Name
histone deacetylase 5
There are no transcripts associated with this gene.

Gene
LSM12
Gene Name
LSM12 homolog
There are no transcripts associated with this gene.

View Full Product Details